Omentin Human Recombinant is produced in E.coli by recombinant DNA technology is a single, polypeptide chain containing 313 amino ac ids and having a molecular mass of 35 kDa. Intelectin is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80533-10
Supplier: Biorbyt
Frequently Bought Together