Order Entry
Spain
ContactUsLinkComponent
FGF 1 Human Recombinant (FGF-1) produced in Sf9 insect cells is a single, glycosylated, polypeptide chain containing 140 amino acids and having a molecular mass of 15803 Dalton. The FGF-a is purified by proprietary chromatographic technique 1 * 2 µG
Catalog # BIRBORB80306-2
Supplier:  Biorbyt
FGF 1 Human Recombinant (FGF-1) produced in Sf9 insect cells is a single, glycosylated, polypeptide chain containing 140 amino acids and having a molecular mass of 15803 Dalton. The FGF-a is purified by proprietary chromatographic technique 1 * 2 µG
Catalog # BIRBORB80306-2
Supplier:  Biorbyt