Order Entry
Spain
ContactUsLinkComponent
Eotaxin also called CCL11 is a CC chemokine that signals through the CCR3 receptor. It is produced by IFN-gamma stimulatedendothelia l cells and TNF-activated monocytes. Eotaxin selectively chemoattracts eosinophils and along Eotaxin-2 and E 1 * 5 µG
Catalog # BIRBORB107706-5
Supplier:  Biorbyt
Eotaxin also called CCL11 is a CC chemokine that signals through the CCR3 receptor. It is produced by IFN-gamma stimulatedendothelia l cells and TNF-activated monocytes. Eotaxin selectively chemoattracts eosinophils and along Eotaxin-2 and E 1 * 5 µG
Catalog # BIRBORB107706-5
Supplier:  Biorbyt
Frequently Bought Together