Order Entry
Spain
ContactUsLinkComponent
Human CCL18 ELISA Kit. Chemokine (C-C motif) ligand 18 (CCL18) is a small cytokine belonging to the CC chemokine family that was pre viously called PARC (pulmonary and activation-regulated chemokine). CCL18 is approximately 60% identical in 1 * 96 items
Catalog # BIRBORB106610-96
Supplier:  Biorbyt
Human CCL18 ELISA Kit. Chemokine (C-C motif) ligand 18 (CCL18) is a small cytokine belonging to the CC chemokine family that was pre viously called PARC (pulmonary and activation-regulated chemokine). CCL18 is approximately 60% identical in 1 * 96 items
Catalog # BIRBORB106610-96
Supplier:  Biorbyt