Order Entry
Spain
Orders LinkContactUsLinkComponent
CF® Dye BCN
CF® Dye BCN
CF®488A BCN, membrane permeability: Impermeant, 490/515 nm, 0,5 mg
Catalog # 92075.
Supplier:   Biotium
CF® Dye BCN
Catalog # 92075.
Supplier:  Biotium
CAS Number:  
CF® Dye BCN
CF® Dye BCN
CF®488A BCN, membrane permeability: Impermeant, 490/515 nm, 0,5 mg
CAS Number:  
Price
601,00 €
Log in for availability
1 * 0,5 mg
Product Details & Documents
CF® Dyes with a BCN group react with azides via a copper-free bioorthogonal reaction to label target molecules for surface staining of live cells or cell extracts. Membrane-permeant CF® Dye BCN conjugates for intracellular labelling reactions are also available.
  • Fluorescent labelling of azide groups on target molecules via copper-free bioorthonal chemistry
  • Alternative to fluorescently labelled DIBO and DBCO
  • Suitable for surface or intracellular labelling of live cells or cell extracts
  • Bright, photostable and water-soluble CF® Dyes are excellent options for fluorescent labelling
  • Available in several CF® Dye options including membrane-permeant forms for intracellular labelling
CF® Dyes BCN (bicyclo[6.1.0]nonyne) derivatives are alternatives to fluorescently labelled DIBO and DBCO. They react with azides to form 1,2,3-triazole by copper-free bioorthogonal 1,3-dipolar Huisgen cycloaddition. Useful for fluorescent surface labelling of live cells or cell extracts, particularly when there are concerns about native protein function loss with copper. Membrane-permeant CF® Dye BCN options are also available for intracellular labelling reactions.
Available CF® Dye BCN derivatives: CF®405S, CF®405M, CF®488A, CF®568, CF®594, CF®640R, CF®647, CF®680, and CF®680R including membrane-permeant CF®440 BCN, CF®500 BCN, and CF®650 BCN. Biotium's next-generation CF® Dyes were designed to be highly water-soluble with advantages in brightness and photostability compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.
Specifications
Specifications
  • Pk:
    0,5 mg
  • Description:
    CF®488A BCN, membrane permeability: Impermeant
  • Ex/Em:
    490/515 nm
Specifications
Recomendaciones
Las recomendaciones se personalizarán según sus preferencias de compra solo si ha dado su consentimiento activando la opción "Mejorar mi experiencia de compra" en la "página de información personal".
De lo contrario, recibirá recomendaciones genéricas.