Order Entry
Spain
ContactUsLinkComponent
971-fp elsd connection kit. required if elsd to be connected to 971-fp. contains flow splitter, communication cable with usb-serial  adaptor, and tubing. 1 * 1 items
Catalog # VARIAX2050-1
Proveedor:  VARIAN
971-fp elsd connection kit. required if elsd to be connected to 971-fp. contains flow splitter, communication cable with usb-serial  adaptor, and tubing. 1 * 1 items
Catalog # VARIAX2050-1
Proveedor:  VARIAN
Frequently Bought Together