Detergent ESD Jontec Destat 5L 7513122 Concentrated liquid neutral detergent, for antistic floors Formulated to maintain and improve ESD performances Can be used with mopping or with autoscrubber To be diluted between 2 and 7% v/v Case of 2 canisters 1 * 2 Can
Catalog # DIVY7513122
Supplier: Diversey
Frequently Bought Together