Temperature indicating strips
88…110 °C
Catalog # SIMETC5030
Supplier: Industrial Physics
Jump to:
Product Details & Documents
These temperature indicating strips are designed for use in electronics, food and pharmaceutical industries, and also in the oven and engine construction businesses. They are particularly useful for indicating temperatures of moving parts.
- Self adhesive foils to measure maximum surface temperatures
- Indicators change colour when marked temperature is reached and the change is irreversible
- Highest temperature remains visible for months
Entrega: Series 5 and Series 8 contains 10 stickers per sheet.
Documents
Specifications
- Seq:03
- Range:88…110 °C