Entrada de pedido
Spain
Orders LinkContactUsLinkComponent
Injection spikes for multidose vials, Mini Spike Plus®
Injection spikes for multidose vials, Mini Spike Plus®
Injection spikes with venting filter and green cap
Catalog # BRAU4550242
Supplier:   B. Braun
Injection spikes for multidose vials, Mini Spike Plus®
Catalog # BRAU4550242
Supplier:  B. Braun
Injection spikes for multidose vials, Mini Spike Plus®
Injection spikes for multidose vials, Mini Spike Plus®
Injection spikes with venting filter and green cap
Price
202,00 €
Log in for availability
1 * 100 UN
Product Details & Documents
Mini Spike Plus withdrawal and injection spikes for multidose vials and multiple withdrawals.
  • Bacteria-proof venting and comfortable handling
  • Higher safety, increased efficiency, high flow rate
  • Effortless and precise penetration even of thick stoppers
Integrated snap cap suitable for one-handed opening and closing with a reliable protection of drugs against microbial contamination during withdrawal.
Documents
Specifications
  • Descripción:
    Injection spikes with venting filter and green cap
  • Seq:
    07