Order Entry
Spain
Orders LinkContactUsLinkComponent
Spectrophotometer with the absorption module, you can make spectrophotometreci chemical dosings, check the beer-lambert's law, find the concentration of a chemical solution and characterize every absorption spectrum. 1 * 1 items
Spectrophotometer with the absorption module, you can make spectrophotometreci chemical dosings, check the beer-lambert's law, find the concentration of a chemical solution and characterize every absorption spectrum. 1 * 1 items
Catalog # DIDAPOF010360
Supplier:   DIDALAB
Spectrophotometer with the absorption module, you can make spectrophotometreci chemical dosings, check the beer-lambert's law, find the concentration of a chemical solution and characterize every absorption spectrum. 1 * 1 items
Catalog # DIDAPOF010360
Supplier:  DIDALAB
Spectrophotometer with the absorption module, you can make spectrophotometreci chemical dosings, check the beer-lambert's law, find the concentration of a chemical solution and characterize every absorption spectrum. 1 * 1 items
Price
1 741,25 €
Iniciar sesión Para disponibilidad
1 * 1 UN
Product Details & Documents