Order Entry
Spain
Orders LinkContactUsLinkComponent
 

Buffer solutions, ready to use, referenced at 25 °C, Certipur®

10,00 (blue), Boric acid, sodium hydroxide, potassium chloride, 4 L
Catalog # 1.99050.4000
   Sustainable Packaging
Supplier:   Merck
Buffer solutions, ready to use, referenced at 25 °C, Certipur®
Catalog # 1.99050.4000
Supplier:  Merck
   Sustainable Packaging
CAS Number:  
Buffer solutions, ready to use, referenced at 25 °C, Certipur®
Buffer solutions, ready to use, referenced at 25 °C, Certipur®

10,00 (blue), Boric acid, sodium hydroxide, potassium chloride, 4 L

CAS Number:  
Price
196,00 €
Log in for availability
1 * 4 L
Product Details & Documents

Materials for the precise calibration and monitoring of pH meters.

  • Traceable to NIST primary standard and PTB - deviation max. ±0,01 (except pH 10 with ±0,02)
  • Larger 4 litre pack sizes in easy to use Titripac® packaging which eliminates contamination issues
  • Colour coded buffers at pH values 4,00, 7,00 and 10,00 are available
  • Reference temperature: +25 °C

All buffer solutions are measured and qualified in a DIN EN ISO 17025/IEC accredited laboratory for pH measurement.

 : Plastic bottles (500 ml) or PE/aluminium Titripac® pack (4 litre)

Specifications

Specifications

  • Pk:
    4 L
  • pH value:
    10,00 (blue)
  • Contents:
    Boric acid, sodium hydroxide, potassium chloride
  • Cat. No.:
    1.99050.4000
  • Environmentally Preferable:
    Sustainable Packaging

Specifications