REDTAQ DNA POLYMERASE CATALYZES 5'-3' SYNTHESIS OF DNA. THE ENZYME HAS BEEN PROVED NOT HAVING THE 3'-5' EXONUCLEASE ACTIVITY. THE EN ZYME HAS PROVEN TO HAVE HIGH AMPLIFICATION YIELD, BE STABLE AT HIGH TEMPARETAURE. ADDED INERT DYE WILL NOT HAVE ANY I 1 * 1.000 EU
Catalog # ABNOD0012-2
Supplier: Abnova
Frequently Bought Together