Order Entry
Spain
ContactUsLinkComponent
600161 GFTB 2000 Hygro-/Thermo-/Barometer Temperature: -25.0 °C ... +70.0 °C Air humidity: 0.0 ... 100.0 % RH (recommended range: 11 ... 90 % RH) Air pressure: 10.0 ... 1100.0 mbar 1 * 1 items
Catalog # GRSG600161
Supplier:  Greisinger
600161 GFTB 2000 Hygro-/Thermo-/Barometer Temperature: -25.0 °C ... +70.0 °C Air humidity: 0.0 ... 100.0 % RH (recommended range: 11 ... 90 % RH) Air pressure: 10.0 ... 1100.0 mbar 1 * 1 items
Catalog # GRSG600161
Supplier:  Greisinger
Frequently Bought Together