Order Entry
Spain
Orders LinkContactUsLinkComponent
Thrombin (biotinylated)

Thrombin (biotinylated)

500 U/g
Catalog # 69672-500UN
Thrombin (biotinylated)
Catalog # 69672-500UN
CAS Number:  
Thrombin (biotinylated)

500 U/g

CAS Number:  
Price
2 970,00 €
Log in for availability
1 * 1 C/U
Product Details & Documents

Biotinylated Thrombin has covalently attached biotin for easy removal of the enzyme from cleavage reactions using immobilised streptavidin. This preparation is tested for activity using the same assay as for unmodified thrombin, and for >99% binding to streptavidin agarose.

Storage temperature: −20 °C.

Specifications

Specifications

  • Pk:
    500 U/g
  • Cat. No.:
    69672-500UN
  • Enzyme name:
    Thrombin (biotinylated)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.