Order Entry
Spain
Orders LinkContactUsLinkComponent
VWR® Q Path® Slide Holders
Slide holders, Translucent, PP, 2
Catalog # 631-1074
Supplier:   VWR Collection
VWR® Q Path® Slide Holders
Catalog # 631-1074
Supplier:  VWR Collection
VWR® Q Path® Slide Holders
VWR® Q Path® Slide Holders
Slide holders, Translucent, PP, 2
Price
504,00 €
Log in for availability
1 * 600 UN
Product Details & Documents
Slide holders designed for applications in clinical cytology.
  • Fast, easy clip closure
  • For two or three slides, depending on the model
  • Modern, user friendly design
Specifications
  • No. of slides:
    2
  • Colour:
    Translucent
  • Description:
    Slide holders
  • Material:
    PP
  • Seq:
    001