Order Entry
Spain
ContactUsLinkComponent
Single position ultra clean base plate. a base plate allows cartridges to be exchanged without introducing. its features and benefit s include - ultra-high capacity high-purity output ensures 99.9999% pure gas quick connect fittings for easy, leak- 1 * 1 items
Catalog # PERCN9306801
Supplier:  PerkinElmer
Single position ultra clean base plate. a base plate allows cartridges to be exchanged without introducing. its features and benefit s include - ultra-high capacity high-purity output ensures 99.9999% pure gas quick connect fittings for easy, leak- 1 * 1 items
Catalog # PERCN9306801
Supplier:  PerkinElmer
Frequently Bought Together