Order Entry
Spain
ContactUsLinkComponent
recombinant/native protein, csnk1g3 (human) recombinant protein, human csnk1g3 (np_004375.2, 1 a.a. - 447 a.a.) full-length recombin ant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5523
Supplier:  Abnova
recombinant/native protein, csnk1g3 (human) recombinant protein, human csnk1g3 (np_004375.2, 1 a.a. - 447 a.a.) full-length recombin ant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5523
Supplier:  Abnova