Order Entry
Spain
Orders LinkContactUsLinkComponent
Inserts
Inserts
E 104/1 Insert suitable for about 200 Test tubes (up to 12×105 mm), mesh width is 8×8 mm, including lid A 13, Ø 6×130mm
Catalog # 141-0338
Supplier:   MIELE
Inserts
Catalog # 141-0338
Supplier:  MIELE
Inserts
Inserts
E 104/1 Insert suitable for about 200 Test tubes (up to 12×105 mm), mesh width is 8×8 mm, including lid A 13, Ø 6×130mm
Price
297,00 €
Log in for availability
1 * 1 UN
Product Details & Documents
Specifications
  • Description:
    E 104/1 Insert suitable for about 200 Test tubes (up to 12×105 mm), mesh width is 8×8 mm, including lid A 13, Ø 6×130mm
  • Seq:
    29