Order Entry
Spain
Orders LinkContactUsLinkComponent
Inserts
E 105/1 Insert suitable for about 200 Test tubes (up to 12×165 mm), mesh width is 9×9 mm, including lid A 13, Ø 6×210mm
Catalog # 141-0339
Supplier:   MIELE
Inserts
Catalog # 141-0339
Supplier:  MIELE
Inserts
Inserts
E 105/1 Insert suitable for about 200 Test tubes (up to 12×165 mm), mesh width is 9×9 mm, including lid A 13, Ø 6×210mm
Price
340,00 €
Log in for availability
1 * 1 UN
Product Details & Documents
Specifications
  • Description:
    E 105/1 Insert suitable for about 200 Test tubes (up to 12×165 mm), mesh width is 9×9 mm, including lid A 13, Ø 6×210mm
  • Seq:
    30