Order Entry
Spain
Orders LinkContactUsLinkComponent
Inserts
Inserts
E 139/1 Insert suitable for about 200 Test tubes (up to 12×200 mm), mesh width is 9×9 mm, including lid A 13, Ø 4×90mm
Catalog # 141-0340
Supplier:   MIELE
Inserts
Catalog # 141-0340
Supplier:  MIELE
Inserts
Inserts
E 139/1 Insert suitable for about 200 Test tubes (up to 12×200 mm), mesh width is 9×9 mm, including lid A 13, Ø 4×90mm
Price
345,00 €
Log in for availability
1 * 1 UN
Product Details & Documents
Specifications
  • Description:
    E 139/1 Insert suitable for about 200 Test tubes (up to 12×200 mm), mesh width is 9×9 mm, including lid A 13, Ø 4×90mm
  • Seq:
    33