Order Entry
Spain
Orders LinkContactUsLinkComponent
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)

Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)

TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer), Glass bottle, 1 L
Catalog # J63677.K2
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)
Catalog # J63677.K2
CAS Number:  
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)

TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer), Glass bottle, 1 L

CAS Number:  
Price
64,80 €
Log in for availability
1 * 1 L
Product Details & Documents

Physical Form: Liquid
MDL No.: MFCD00236357

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer)
  • Cat. No.:
    J63677.K2
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00236357

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.