TRIS-glycine native sample buffer (4X), pH 8,6
TRIS-Glycine Native Sample Buffer (4X), 8,6, 50 mL
Supplier: Thermo Fisher Scientific
TRIS-glycine native sample buffer (4X), pH 8,6
Catalog # J61864.AD
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Physical Form: Liquid
Appearance: Colourless to yellow
MDL No.: MFCD00236360
Applications: For preparation of protein samples for native gel electrophoresis
Documents
Specifications
Specifications
- Pk:50 mL
- Description:TRIS-Glycine Native Sample Buffer (4X)
- pH value:8,6
- Cat. No.:J61864.AD
- Physical form:Liquid
- MDL No.:MFCD00236360
- Appearance:Colourless to yellow
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



