Order Entry
Spain
Orders LinkContactUsLinkComponent
 

Buffer, ACES, 0,5 M solution

ACES, 0,5M buffer solution, pH 7,0, Glass bottle, 250 mL
Catalog # J60755.AK
Buffer, ACES, 0,5 M solution
Catalog # J60755.AK
CAS Number:  
Buffer, ACES, 0,5 M solution
Buffer, ACES, 0,5 M solution

ACES, 0,5M buffer solution, pH 7,0, Glass bottle, 250 mL

CAS Number:  
Price
103,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

CAS No.: 7365-82-4
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00008030

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Description:
    ACES, 0,5M buffer solution, pH 7,0
  • Cat. No.:
    J60755.AK
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00008030
  • Appearance:
    Colourless
  • CAS No.:
    7365-82-4

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.