Order Entry
Spain
ContactUsLinkComponent
Stirrer Bar with champhered magnet core, 75mmL X 13mm DIA. Each stirrer bar will be engraved with the Job Number. Product will be Fully Traceable to production and material batch numbers. The product batch will have a 100% Visual inspection using 2 x 1 * 1 items
Catalog # COWIFT.001.575.NOV
Supplier:  COWIE
Stirrer Bar with champhered magnet core, 75mmL X 13mm DIA. Each stirrer bar will be engraved with the Job Number. Product will be Fully Traceable to production and material batch numbers. The product batch will have a 100% Visual inspection using 2 x 1 * 1 items
Catalog # COWIFT.001.575.NOV
Supplier:  COWIE
Frequently Bought Together