Order Entry
Spain
Orders LinkContactUsLinkComponent
Steps, safety

Steps, safety

Safety steps with 2 steps, aluminium chequer plate, 460 mm, —
Catalog # WAKU220C
Supplier:   WAKU GERATE
Steps, safety
Catalog # WAKU220C
Supplier:  WAKU GERATE
Steps, safety
Steps, safety

Safety steps with 2 steps, aluminium chequer plate, 460 mm, —

Price
320,00 €
Log in for availability
1 * 1 UN
Product Details & Documents

Light, aluminium safety steps. Strong double rails with robust, tension and pressure resistant bracings for increased stability. Platform height corresponds to height of top step.

  • Anti-slip plastic feet
  • The large treads (36×23 cm) are covered with a non-slip rubber lining.

TÜV tested (DIN 4566 and 4567).

Certifications: TÜV tested (DIN 4566 and 4567).

Specifications
  • Description:
    Safety steps with 2 steps, aluminium chequer plate
  • Height:
    460 mm
  • Weight: