Order Entry
Spain
ContactUsLinkComponent
Dish evaporating 95x55mm glass 170ml made of borosilicate glass 3.3 in compliance with din 12 336 very good chemical resistance mini mal thermal expansion high resistance to thermal shocks with fused rim+spout 1 * 10 items
Catalog # MARI4210309
Supplier:  MARIENFELD
Dish evaporating 95x55mm glass 170ml made of borosilicate glass 3.3 in compliance with din 12 336 very good chemical resistance mini mal thermal expansion high resistance to thermal shocks with fused rim+spout 1 * 10 items
Catalog # MARI4210309
Supplier:  MARIENFELD
Frequently Bought Together