Order Entry
Spain
Orders LinkContactUsLinkComponent
 

Sulfate standard solution

Sulfate standard solution, 500 ml (NIST), 2500 mg/l as SO42-, 1
Catalog # HACH1425249
Supplier:   Hach
Sulfate standard solution
Catalog # HACH1425249
Supplier:  Hach
CAS Number:  
Sulfate standard solution
Sulfate standard solution

Sulfate standard solution, 500 ml (NIST), 2500 mg/l as SO42-, 1

CAS Number:  
Price
50,60 €
Log in for availability
1 * 1 UN
Product Details & Documents

Standard solution for accuracy checks in sulfate determinations.

Specifications

Specifications

  • Concentration:
    2500 mg/l as SO<sub>4</sub><sup>2-</sup>
  • Pk:
    1
  • Description:
    Sulfate standard solution, 500 ml (NIST)

Specifications