Order Entry
Spain
Orders LinkContactUsLinkComponent
Tween® 80 (Polysorbate)

Tween® 80 (Polysorbate)

Glass bottle, 500 g
Catalog # TCIAT0546-500G
Irritant
Supplier:   TCI
Tween® 80 (Polysorbate)
Catalog # TCIAT0546-500G
Supplier:  TCI
CAS Number:  9005-65-6
Tween® 80 (Polysorbate)

Glass bottle, 500 g

CAS Number:  9005-65-6
Price
29,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Merck Index:14,07582
  • Boiling Pt:? 100 °C (1013 hPa)
  • Melting Pt:–21 °C
  • Density:1,062 g/cm³ (20 °C)
  • Flash Pt:295°C
  • Storage Temperature:Ambient
  • EINECS:500-019-9
Specifications

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green clear liquid
  • Saponification Value:
    45.0 to 55.0
  • Water:
    max. 4.0 %
  • Acid Value:
    max. 2.0

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g

Specifications