Tween® 80 (Polysorbate)
Glass bottle, 25 g
Catalog # TCIAT0546-25G
Supplier: TCI
Jump to:
Product Details & Documents
- CAS Number:9005-65-6
- MDL Number:MFCD00082107
- Merck Index:14,07582
- Boiling Pt:? 100 °C (1013 hPa)
- Melting Pt:–21 °C
- Density:1,062 g/cm³ (20 °C)
- Flash Pt:295°C
- Storage Temperature:Ambient
- EINECS:500-019-9
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Saponification Value:45.0 to 55.0
- Water:max. 4.0 %
- Acid Value:max. 2.0
Specifications
- Pack type:Glass bottle
- Pk:25 g



