Order Entry
Spain
Orders LinkContactUsLinkComponent
Temperature indicating strips
Temperature indicating strips
143…166 °C
Catalog # SIMETC5040
Supplier:   Industrial Physics
Temperature indicating strips
Catalog # SIMETC5040
Supplier:  Industrial Physics
Temperature indicating strips
Temperature indicating strips
143…166 °C
Price
36,70 €
Log in for availability
1 * 10 UN
Product Details & Documents
These temperature indicating strips are designed for use in electronics, food and pharmaceutical industries, and also in the oven and engine construction businesses. They are particularly useful for indicating temperatures of moving parts.
  • Self adhesive foils to measure maximum surface temperatures
  • Indicators change colour when marked temperature is reached and the change is irreversible
  • Highest temperature remains visible for months
Delivery information: Series 5 and Series 8 contains 10 stickers per sheet.
Specifications
  • Seq:
    05
  • Range:
    143…166 °C