Order Entry
Spain
ContactUsLinkComponent
ChemiFast : Substrate specifically configured for use with cooled CCD based imaging systems which is optimised for high sensitivity and long signal duration. High emission intensity of up to 24 hours is possible. 1 * 1 items
Catalog # SYNOCHFAST-100
ChemiFast : Substrate specifically configured for use with cooled CCD based imaging systems which is optimised for high sensitivity and long signal duration. High emission intensity of up to 24 hours is possible. 1 * 1 items
Catalog # SYNOCHFAST-100
Frequently Bought Together