Order Entry
Spain
Orders LinkContactUsLinkComponent
Ethyl oleate

Ethyl oleate

Glass bottle for solids, 1 g
Catalog # CAYM10008201-1
Supplier:   Cayman Chemical
Ethyl oleate
Catalog # CAYM10008201-1
Supplier:  Cayman Chemical
CAS Number:  111-62-6
Ethyl oleate

Glass bottle for solids, 1 g

CAS Number:  111-62-6
Price
59,40 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Etilo oleato
  • CAS Number:111-62-6
  • Formula:C₂₀H₃₈O₂
  • MDL Number:MFCD00009579
  • Molecular Weight:310.52 g/mol
  • Merck Index:12,06965
  • Boiling Pt:386 °C (1013 hPa)
  • Melting Pt:–32 °C
  • Density:0,87 g/cm³ (20 °C)
  • Flash Pt:174 °C
  • Storage Temperature:Ambiente
  • EINECS:203-889-5
Specifications

Specifications

  • Tipo de env.:
    Glass bottle for solids
  • Env:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.