Order Entry
Spain
Orders LinkContactUsLinkComponent
Test kits, chlorine (DPD), CHECKIT®
Chlorine test kit, DPD (free + total), powder pack, 0 - 3,5 mg/l Cl2, 50 Tests
  LOVI147052
 :   Lovibond
Test kits, chlorine (DPD), CHECKIT®
  LOVI147052
 :  Lovibond
 :  
Test kits, chlorine (DPD), CHECKIT®
Test kits, chlorine (DPD), CHECKIT®
Chlorine test kit, DPD (free + total), powder pack, 0 - 3,5 mg/l Cl2, 50 Tests
 :  
108,00 €
1 * 50 Tests
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work. Supplied with a generous number of different colour scales, it provides the basis for a comprehensive, easy-to use colorimetric analysis system.
  • Enables the use of large path lengths
  • The mirror optics makes use of the view through the entire length of the cell
  • Easy operation and exact reagent dosing
  • Tablet reagents have minimum guaranteed shelf life of 5/10 years
The operating instructions provide a step-by-step explanation of conducting the water test to achieve reliable and accurate measurements in the minimum of time.
CHECKIT® disc contains a continuous colour scale which makes it possible to achieve an exact colour match between the colour standard and the sample. These discs are specially manufactured in selected materials to remain colour-stability over a long period and guarantee reliable, reproducible measurement results.
 : Test kit includes CHECKIT® discs, cells, stirring rod and AQUALYTIC® tablet reagents (for 30 tests) for the desired test supplied in a sturdy plastic case.
  • Pk:
    50 Tests
  • Description:
    Chlorine test kit, DPD (free + total), powder pack
  • Range:
    0 - 3,5 mg/l Cl<sub>2</sub>
Accessories
  •  
    Description
    Pk
  • LOVI145000
    CHECKIT® Comparator, path length 13,5 mm
    1
    1 * 1 UN - 20,40 €
  •  
    Description
    Pk
  • LOVI145500
    CHECKIT® plastic cells, pack of 10, with lid, 10 ml, 13,5 mm path length
    10
    1 * 10 UN - 46,30 €
  •  
    Description
    Pk
  • LOVI145505
    CHECKIT® plastic cells, pack of 5, with lid, 10 ml, 13,5 mm path length
    5
    1 * 5 UN - 27,40 €
  •  
    Description
    Pk
  • LOVI145510
    CHECKIT® plastic cells, pack of 100, with lid, 10 ml, 13,5 mm path length
    100
    1 * 100 UN - 383,00 €