Benzonase®
100,000 U/vial, Plastic bottle, 1
Supplier: MERCK PRODUCTION CHEMICALS
Jump to:
Product Details & Documents
Benzonase® is a highly active nuclease that degrades DNA and RNA to short oligonucleotides. It is used for the elimination of interfering DNA/RNA, for example at protein isolation, or before electrophoresis or chromatography.
Delivery information: Aluminium tube containing 500000 Units and Plastic bottle containing 100000 units.
Documents
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:1
- Pk Info:100,000 U/vial
- Cat. No.:1.01654.0001
- Enzyme name:Benzonase®