Order Entry
Spain
Orders LinkContactUsLinkComponent
Benzonase®
Benzonase®
500,000 U/vial, Aluminium tube, 1
Catalog # 1.01656.0001
Benzonase®
Catalog # 1.01656.0001
CAS Number:  
Benzonase®
500,000 U/vial, Aluminium tube, 1
CAS Number:  
Price
4 160,00 €
Log in for availability
1 * 1 UN
Product Details & Documents
Benzonase® is a highly active nuclease that degrades DNA and RNA to short oligonucleotides. It is used for the elimination of interfering DNA/RNA, for example at protein isolation, or before electrophoresis or chromatography.
Entrega: Aluminium tube containing 500000 Units and Plastic bottle containing 100000 units.
Specifications
Specifications
  • Pack type:
    Aluminium tube
  • Pk:
    1
  • Pk Info:
    500,000 U/vial
  • Cat. No.:
    1.01656.0001
  • Enzyme name:
    Benzonase®