Order Entry
Spain
Orders LinkContactUsLinkComponent
Test kits, chlorine (DPD), CHECKIT®

Test kits, chlorine (DPD), CHECKIT®

Chlorine test kit, DPD (free), powder pack, 0 - 3,5 mg/l Cl2, 1 KIT
Catalog # LOVI147050
Supplier:   Lovibond
Test kits, chlorine (DPD), CHECKIT®
Catalog # LOVI147050
Supplier:  Lovibond
CAS Number:  
Test kits, chlorine (DPD), CHECKIT®
Test kits, chlorine (DPD), CHECKIT®

Chlorine test kit, DPD (free), powder pack, 0 - 3,5 mg/l Cl2, 1 KIT

CAS Number:  
Price
108,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work. Supplied with a generous number of different colour scales, it provides the basis for a comprehensive, easy-to use colorimetric analysis system.

  • Enables the use of large path lengths
  • The mirror optics makes use of the view through the entire length of the cell
  • Easy operation and exact reagent dosing
  • Tablet reagents have minimum guaranteed shelf life of 5/10 years

The operating instructions provide a step-by-step explanation of conducting the water test to achieve reliable and accurate measurements in the minimum of time.

CHECKIT® disc contains a continuous colour scale which makes it possible to achieve an exact colour match between the colour standard and the sample. These discs are specially manufactured in selected materials to remain colour-stability over a long period and guarantee reliable, reproducible measurement results.

Delivery information: Test kit includes CHECKIT® discs, cells, stirring rod and AQUALYTIC® tablet reagents (for 30 tests) for the desired test supplied in a sturdy plastic case.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Chlorine test kit, DPD (free), powder pack
  • Range:
    0 - 3,5 mg/l Cl<sub>2</sub>

Specifications

Accessories

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • CHECKIT® Comparator, path length 13,5 mm
    CHECKIT® Comparator, path length 13,5 mm
    LOVI145000
    1
    Log in for availability
    1 * 1 UN - 20,40 €

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • CHECKIT® plastic cells, pack of 10, with lid, 10 ml, 13,5 mm path length
    CHECKIT® plastic cells, pack of 10, with lid, 10 ml, 13,5 mm path length
    LOVI145500
    10
    Log in for availability
    1 * 10 UN - 46,30 €

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • CHECKIT® plastic cells, pack of 5, with lid, 10 ml, 13,5 mm path length
    CHECKIT® plastic cells, pack of 5, with lid, 10 ml, 13,5 mm path length
    LOVI145505
    5
    Log in for availability
    1 * 5 UN - 27,40 €

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • CHECKIT® plastic cells, pack of 100, with lid, 10 ml, 13,5 mm path length
    CHECKIT® plastic cells, pack of 100, with lid, 10 ml, 13,5 mm path length
    LOVI145510
    100
    Log in for availability
    1 * 100 UN - 383,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.