Order Entry
Spain
ContactUsLinkComponent
Gel Documentation System, designed specifically for Chemiluminesce work. With 1.4 Mega Pixel camera and 2 copies of GeneTools analysis software 1 * 1 items
Catalog # 733-2307
Gel Documentation System, designed specifically for Chemiluminesce work. With 1.4 Mega Pixel camera and 2 copies of GeneTools analysis software 1 * 1 items
Catalog # 733-2307
Frequently Bought Together