Order Entry
Spain
ContactUsLinkComponent
STIRRED BATH/CIRCULATOR, OPTIMA TXF200 PROGRAMMABLE IMMERSION THERMOSTAT + ST5 STAINLESS STEEL TANK, 5L CAPACITY. TEMP RANGE AMBIENT +5 TO 200C. INCL. PUMP. 1 * 1 items
Catalog # 461-0720
Supplier:  GRANT INSTRUMENTS
STIRRED BATH/CIRCULATOR, OPTIMA TXF200 PROGRAMMABLE IMMERSION THERMOSTAT + ST5 STAINLESS STEEL TANK, 5L CAPACITY. TEMP RANGE AMBIENT +5 TO 200C. INCL. PUMP. 1 * 1 items
Catalog # 461-0720
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together