Order Entry
Spain
ContactUsLinkComponent
STIRRED BATH, OPTIMA T100 IMMERSION THERMOSTAT + ST26 STAINLESS STEEL TANK, 26L CAPACITY, INCL DRAIN TAP. TEMP RANGE AMBIENT +5 TO 1 00C. 1 * 1 items
Catalog # GRANT100-ST26EU
Supplier:  GRANT INSTRUMENTS
STIRRED BATH, OPTIMA T100 IMMERSION THERMOSTAT + ST26 STAINLESS STEEL TANK, 26L CAPACITY, INCL DRAIN TAP. TEMP RANGE AMBIENT +5 TO 1 00C. 1 * 1 items
Catalog # GRANT100-ST26EU
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together