Order Entry
Spain
ContactUsLinkComponent
Jetpei transfection reagent is a linear polyethylenimine derivative, providing highly effective and reproducible gene delivery.it is particularly well suited for high throughput screening. 1 * 10 ml
Catalog # 101B-010
Jetpei transfection reagent is a linear polyethylenimine derivative, providing highly effective and reproducible gene delivery.it is particularly well suited for high throughput screening. 1 * 10 ml
Catalog # 101B-010