Wright's stain solution for microscopy, Sigma-Aldrich®
250 mL
45253-250ML-F
Wright's stain solution for microscopy, Sigma-Aldrich®
45253-250ML-F
: 68988-92-1
Wright Stain solution, for microscopy (Eosin Methylene blue solution according to Wright; Wright's eosin methylene blue)
Wright Stain is a modified Romanowsky stain commonly used for the cellular elements of blood. Wright′s stain is a mixture of eosin Y with polychromed (oxidized and partly demethylated) methylene blue and therefore termed polychrome stains. The hematological stain is intended for differentially staining peripheral blood and bone marrow films.
: Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue
- :68988-92-1
- :MFCD00005038
- :0,80 g/ml (20 °C)
- :1230
- :3,II
- :M-Clarity™ quality level = MQ100
- :11 °C
- Appearance (colour):Blue to very dark blue
- Appearance (form):Liquid
- Thin layer chromatogr.:Corresponds to standard
- Density D20/4:0.785 - 0.814
- Wavelength (1) (UV):635 - 650 nm
- Extinction (1%/1 cm) (1) (UV):--
- Wavelength (2) (UV):512 - 527 nm
- Extinction (1%/1 cm) (2) (UV):--
- Solvent (UV):Meoh
- Expiry period:30 months
- Pk:250 mL
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



