Order Entry
Spain
Orders LinkContactUsLinkComponent
pH buffer solutions

pH buffer solutions

Technical buffer solution, 6×250 ml, 2,00
Catalog # METT51350016
Supplier:   Mettler - Toledo
pH buffer solutions
Catalog # METT51350016
Supplier:  Mettler - Toledo
CAS Number:  
pH buffer solutions
pH buffer solutions

Technical buffer solution, 6×250 ml, 2,00

CAS Number:  
Price
91,60 €
Log in for availability
1 * 6 UN
Product Details & Documents
Specifications

Specifications

  • Description:
    Technical buffer solution, 6×250 ml
  • pH value:
    2,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.