Order Entry
Spain
Orders LinkContactUsLinkComponent
pH buffer solutions

pH buffer solutions

Technical buffer solution, 6×250 ml, 7,00
Catalog # METT51350020
Supplier:   Mettler - Toledo
pH buffer solutions
Catalog # METT51350020
Supplier:  Mettler - Toledo
CAS Number:  
pH buffer solutions
pH buffer solutions

Technical buffer solution, 6×250 ml, 7,00

CAS Number:  
Price
91,10 €
Log in for availability
1 * 6 UN
Product Details & Documents
Specifications

Specifications

  • Description:
    Technical buffer solution, 6×250 ml
  • pH value:
    7,00

Specifications