Order Entry
Spain
Orders LinkContactUsLinkComponent
HPLC columns, PLRP-S

HPLC columns, PLRP-S

PLRP-S, 100 Å, 3 µm, 2,10 mm, 50 mm
Catalog # 568-0820
Supplier:   VARIAN
HPLC columns, PLRP-S
Catalog # 568-0820
Supplier:  VARIAN
HPLC columns, PLRP-S
HPLC columns, PLRP-S

PLRP-S, 100 Å, 3 µm, 2,10 mm, 50 mm

Price
779,53 €
Log in for availability
1 * 1 UN
Product Details & Documents

The PLRP-S family of products consists of a range of pore sizes and particle sizes, all with identical chemistry and fundamental adsorptive characteristics. The particles are inherently hydrophobic so there is no bonded phase, alkyl ligand required for reversed-phase separations. This gives a highly reproducible material that is free from silanols and heavy metal ions. Within the extensive product range are columns suitable for analytical separations and preparative purifications.

  • Contain durable and resilient particles that deliver reproducible results over longer lifetimes
  • Thermally and chemically stable
  • Used in bioscience, chemical, clinical research, energy, environmental, food and agriculture, material science and pharmaceutical industries

Buffer content: unlimited.

Organic modifier: 1 to 100%.

Maximum pressure: 5 to 8 μm: 210 bar; 3 μm: 300 bar.

Applications:
100 Å - Small molecules/synthetic biomolecules
300 Å - Recombinant peptides/proteins
1000 Å - Large proteins
4000 Å - DNA/high speed

Specifications
  • Ø int.:
    2,10 mm
  • Particle Size:
    3 µm
  • Pore size:
    100 Å
  • Length:
    50 mm
  • Seq:
    01
  • Phase:
    PLRP-S
  • Cat. No.:
    568-0820
  • USP code:
    L21
  • Temperature limit:
    200 °C
  • pH range:
    1 - 14
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Guard cartridge holder for 3,0×5,0 mm cartridges
    Guard cartridge holder for 3,0×5,0 mm cartridges
    VARIPL1310-0016
    Log in for availability
    1 * 1 UN - 366,23 €

Product Information

  • Catalog #
    Availability
    Price
  • PLRP-S guard cartridges, for 5×3 mm columns
    PLRP-S guard cartridges, for 5×3 mm columns
    VARIPL1612-1801
    Log in for availability
    1 * 1 UN - 254,73 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.