Order Entry
Spain
Orders LinkContactUsLinkComponent
 

Thrombin (biotinylated)

1.000 U/g
Catalog # 69672-1000UN
Thrombin (biotinylated)
Catalog # 69672-1000UN
CAS Number:  
Thrombin (biotinylated)

1.000 U/g

CAS Number:  
Price
4 810,00 €
Log in for availability
1 * 1 C/U
Product Details & Documents

Biotinylated Thrombin has covalently attached biotin for easy removal of the enzyme from cleavage reactions using immobilised streptavidin. This preparation is tested for activity using the same assay as for unmodified thrombin, and for >99% binding to streptavidin agarose.

Storage temperature: −20 °C.

Specifications

Specifications

  • Pk:
    1.000 U/g
  • Cat. No.:
    69672-1000UN
  • Enzyme name:
    Thrombin

Specifications