Order Entry
Spain
Orders LinkContactUsLinkComponent
QuEChERS extraction products
QuEChERS extraction products
Column with 250 mg graphatised carbon on top, 500 mg PSA on bottom, separated by a PTFE frit, 6 ml
Catalog # 549-0764
QuEChERS extraction products
Catalog # 549-0764
QuEChERS extraction products
QuEChERS extraction products
Column with 250 mg graphatised carbon on top, 500 mg PSA on bottom, separated by a PTFE frit, 6 ml
Price
315,00 €
Log in for availability
1 * 30 UN
Product Details & Documents
The QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) technique is increasingly becoming the technique of choice for the extraction of pesticide residues in fruit, vegetables and other foods. The QuEChERS method offers the advantage of high recoveries, accurate results and high sample throughput, combined with the associated cost savings with low solvent and labour requirements. Yet the procedure is robust and rugged.
  • Simple manual procedure
  • Determine wide range of pesticide types
  • Less solvent usage than conventional techniques
  • High recoveries, accurate results and high sample throughput
Specifications
  • Capacity:
    6 ml
  • Description:
    Column with 250 mg graphatised carbon on top, 500 mg PSA on bottom, separated by a PTFE frit
  • Seq:
    19