Order Entry
Spain
Orders LinkContactUsLinkComponent
Transfer pipettes
Transfer pipettes
Extended fine tip, 1 ml, -
Catalog # 612-4857
Supplier:   DELTALAB
Transfer pipettes
Catalog # 612-4857
Supplier:  DELTALAB
Transfer pipettes
Transfer pipettes
Extended fine tip, 1 ml, -
Price
142,00 €
Log in for availability
1 * 400 UN
Product Details & Documents
PE, disposable.
  • Fine tip
Documents
Specifications
  • Sterile:
    -
  • Capacity:
    1 ml
  • Description:
    Extended fine tip
  • Seq:
    01