MultiCam 310C colour camera: 16 File Bit Depth (A/D) digital camera with mount, 1600 x 1200 chip (2 Mp). 12.5-75mm/f1.2 MOTORISED zoom lens, close up diopter, USB card and cable. GelDoc-It TS light-tight darkroom with built-in PC, 15 LCD touch scree 1 * 1 items
Catalog # UVPA97-0556-02
Supplier: UVP ULTRA VIOLET PRODUCTS
Frequently Bought Together