Order Entry
Spain
Orders LinkContactUsLinkComponent
Adapters with external thread

Adapters with external thread

Adapter M with 4,0 mm O.Ø nipple
Catalog # PERCB0507919
Supplier:   PerkinElmer
Adapters with external thread
Catalog # PERCB0507919
Supplier:  PerkinElmer
Adapters with external thread

Adapter M with 4,0 mm O.Ø nipple

Price
51,60 €
Log in for availability
1 * 1 UN
Product Details & Documents

Adapters with external screw thread (6,4 mm) for FIMS-100, FIMS-400, FIAS-100 or FIAS-400 flow injection system.

Specifications
  • Description:
    Adapter M with 4,0 mm O.Ø nipple
  • Seq:
    03