L(+)-Glutamic acid
Glass bottle for solids, 1 kg
Catalog # A1704.1000
Supplier: PanReac AppliChem
Jump to:
Product Details & Documents
Synonyms: (S)-(+)-2-Aminopentanedioic acid, L(+)-1-Aminopropane-1,3-dicarboxylic acid, (S)-(+)-α-Glutamic acid, H-Glu-OH, L-Glutamic acid, L(+)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid, L(+)-α-Glutamic acid
- CAS Number:56-86-0
- Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
- MDL Number:MFCD00002634
- Molecular Weight:147.13 g/mol
- Merck Index:13,04482
- Boiling Pt:325 °C (1013 hPa)
- Melting Pt:160 °C
- Density:1.538 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-293-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



